Cryoboxes, 133x133 mm
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HS2860A HS2860B HS2860H HS2860F HS2860E HS2860G
HEATHS2860AEA 53.2 EUR
HEATHS2860A HEATHS2860B HEATHS2860H HEATHS2860F HEATHS2860E HEATHS2860G
Cryoboxes, 133x133 mm
Dozen Dozen voor cryobuisjes
Cryoboxes, made of cardboard, for storage of small samples and tubes.
  • Store in LN₂, or in mechanical freezers
  • Available in two different heights with different dividers (to be ordered separately)
  • Colors allow for easy identification and sorting purposes
  • Bottom vent holes to allow quicker circulation and removal of cold air, vapor or LN₂
Leveringsinformatie: Boxes are supplied with lid.