Flasks, round bottom with four necks and standard ground joints, side necks angled (7°)
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03347358 03347458 03347449 03347470 03347349
LENZ03347358EA 112 EUR
LENZ03347358 LENZ03347458 LENZ03347449 LENZ03347470 LENZ03347349
Flasks, round bottom with four necks and standard ground joints, side necks angled (7°)
Kolven Rondbodemkolven
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certificaten: DIN 12392