Flasks, round bottom with four necks and standard ground joints, side necks angled (15°)
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03348449 03348458
LENZ03348449EA 114 EUR
LENZ03348449 LENZ03348458
Flasks, round bottom with four necks and standard ground joints, side necks angled (15°)
Kolven Rondbodemkolven
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.