Boron standard solution, 1000 mg/L B in water (from B(OH₃)), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39147.AD 39147.AE 39147.AP
39147.ADEA 54.1 EUR
39147.AD 39147.AE 39147.AP
Boron standard solution, 1000 mg/L B in water (from B(OH₃)), standard for ICP, Specpure®
Standaarden Kationische standaarden Borium standaarden
Molecular Formula: H₃BO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00134034
Certificaten: Traceable to NIST.