Arsenic standard solution, 10000 mg/L As in 5% nitric acid (from As), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14369.AD 14369.AE 14369.AP
14369.ADEA 86.3 EUR
14369.AD 14369.AE 14369.AP
Arsenic standard solution, 10000 mg/L As in 5% nitric acid (from As), standard for ICP, Specpure®
Standaarden Kationische standaarden Arseen standaarden
Molecular Formula: As in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309
Certificaten: Traceable to NIST