Arsenic standard solution, 1000 mg/L in 5% nitric acid (from As), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33559.AE
33559.AP
33559.AEEA
39.5
EUR
33559.AE
33559.AP
Arsenic standard solution, 1000 mg/L in 5% nitric acid (from As), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Arseen standaarden
Molecular Formula: As in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309
Certificaten: Traceable to NIST