Barium standard solution, 10000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14423.AE
14423.AD
14423.AP
14423.AEEA
131
EUR
14423.AE
14423.AD
14423.AP
Barium standard solution, 10000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Barium standaarden
Molecular Formula: Ba(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134031
Certificaten: Traceable to NIST