Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13876.AD 13876.AE 13876.AP
13876.ADEA 51.5 EUR
13876.AD 13876.AE 13876.AP
Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Standaarden Kationische standaarden Barium standaarden
Molecular Formula: Ba(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134031
Certificaten: Traceable to NIST