Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88052.AE 88052.AP
88052.AEEA 38.7 EUR
88052.AE 88052.AP
Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for AAS, Specpure®
Standaarden Kationische standaarden Barium standaarden
Molecular Formula: Ba(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134031
Certificaten: Traceable to NIST