Lead standard solution, 1000 mg/L Pb in 5% nitric acid (from Pb), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88075.AE
88075.AP
88075.AEEA
40
EUR
88075.AE
88075.AP
Lead standard solution, 1000 mg/L Pb in 5% nitric acid (from Pb), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Lood standaarden
Molecular Formula: Pb in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134050
Certificaten: Traceable to NIST