Cadmium plasma standard solutions, for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13813.AD 13813.AE 13813.AP 14424.AE 14424.AP
13813.ADEA 53.7 EUR
13813.AD 13813.AE 13813.AP 14424.AE 14424.AP
Cadmium plasma standard solutions, for ICP, Specpure®
Standaarden Kationische standaarden Cadmium standaarden
Molecular Formula: Cd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914
Certificaten: Traceable to NIST