Cadmium standard solution, 1000 mg/L Cd in 5% nitric acid (from Cd), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88056.AE 88056.AP
88056.AEEA 40.5 EUR
88056.AE 88056.AP
Cadmium standard solution, 1000 mg/L Cd in 5% nitric acid (from Cd), standard for AAS, Specpure®
Standaarden Kationische standaarden Cadmium standaarden
Molecular Formula: Cd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914
Certificaten: Traceable to NIST