Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13822.AD
13822.AE
13822.AP
13822.ADEA
52.4
EUR
13822.AD
13822.AE
13822.AP
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Bismut standaarden
Molecular Formula: Bi in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033
Certificaten: Traceable to NIST