Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88054.AE 88054.AP
88054.AEEA 39.2 EUR
88054.AE 88054.AP
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for AAS, Specpure®
Standaarden Kationische standaarden Bismut standaarden
Molecular Formula: Bi in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033
Certificaten: Traceable to NIST