Ethyl cyanoformate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C1266-25G
C1266-5G
TCIAC1266-25GEA
86.9
EUR
TCIAC1266-25G
TCIAC1266-5G
Ethyl cyanoformate ≥98.0% (by GC)
Ethyl cyanoformate
Formule:
C₄H₅NO₂ Kookpunt: 116°C Vlampunt: 24°C |
MDL Nummer:
MFCD00001836 CAS nummer: 623-49-4 EINECS: 210-797-9 UN: 3275 ADR: 6.1,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...