Dextran, M.W. ~ 40,000
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A2249.0500EA 219 EUR
A2249.0500
Dextran, M.W. ~ 40,000
Dextran
Formule: (C₆H₁₀O₅)n
MDL Nummer: MFCD00130935
CAS nummer: 9004-54-0
EINECS: 232-677-5