2,5-Piperazinedione

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4030536.0025 4030536.0100
BACE4030536.0025EA 0 EUR
BACE4030536.0025 BACE4030536.0100
2,5-Piperazinedione
2,5-Piperazinedione
Formule: C₄H₆N₂O₂
MDL Nummer: MFCD00006009
CAS nummer: 106-57-0
EINECS: 203-411-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR