Antimony standard solution, 1000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (from Sb), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41682.AD
41682.AE
41682.AP
41682.ADEA
54.9
EUR
41682.AD
41682.AE
41682.AP
Antimony standard solution, 1000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (from Sb), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Antimonium standaarden
Molecular Formula: Sb in H₂O/tartaric acid/tr. HNO₃
Physical Form: Liquid
MDL No.: MFCD00134030
Certificaten: Traceable to NIST