4-(4-Chlorophenyl)piperidin-4-ol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-(4-Chlorophenyl)-4-hydroxypiperidine

BTB09284.1 BTB09284EA
MAYBBTB09284.1EA 0 EUR
MAYBBTB09284.1 MAYBBTB09284EA
4-(4-Chlorophenyl)piperidin-4-ol
4-(4-Chlorophenyl)piperidin-4-ol
Formule: C₁₁H₁₄ClNO
MW: 211.69 g/mol
MDL Nummer: MFCD00006001
CAS nummer: 39512-49-7
EINECS: 254-479-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR