Pyruvaldehyde (methylglyoxal) 40% in aqueous solution
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6399.1 6399.2
ROTH6399.1EA 0 EUR
ROTH6399.1 ROTH6399.2
Pyruvaldehyde (methylglyoxal) 40% in aqueous solution
Pyruvaldehyde (methylglyoxal)
Formule: C₃H₄O₂
MDL Nummer: MFCD00006960
CAS nummer: 78-98-8
EINECS: 201-164-8