9-Fluorenylmethyl chloroformate (Fmoc chloride) for synthesis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl
M-Clarity™ quality level = MQ200
8.18203.0025 8.18203.8500 8.18203.0005
8.18203.0025EA 288 EUR
8.18203.0025 8.18203.8500 8.18203.0005
9-Fluorenylmethyl chloroformate (Fmoc chloride) for synthesis
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formule: C₁₅H₁₁ClO₂
MW: 258.7 g/mol
MDL Nummer: MFCD00001138
CAS nummer: 28920-43-6
EINECS: 249-313-6
UN: 1759
ADR: 8 II