4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (4E)-4-(Ethoxymethylene)-2-phenyl-1,3-oxazol-5(4H)-one, 4-Ethoxymethylene-2-phenyloxazolin-5-one
E0753-10G E0753-1G E0753-5G
E0753-10GEA 381 EUR
E0753-10G E0753-1G E0753-5G
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one, Supelco®
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one
Formule: C₁₂H₁₁NO₃
MDL Nummer: MFCD03225480
CAS nummer: 15646-46-5
EINECS: 239-713-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cf2192371c49f8bc7e99346d06f1512fa402842&vid=9de6ab4da47775913e140f98c5c84b041bfe4ad5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de6d0611eb52d88c51edc62feb6c815745ac6c52&vid=1b581dbf4f16edaeddde177f86f3b4ca29645a25&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb481a00e2d8492cd891c269458e6076640dfc10&vid=2a52ed9dbd039f5add3d7a17aadc37a29e0f24f1&prid=web&clid=SAPDAM