Temperature probes, for agressive liquids, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1750-10 P1750-25 P1750-15 P1750-20 P1750-30
BOHLP1750-10EA 257 EUR
BOHLP1750-10 BOHLP1750-25 BOHLP1750-15 BOHLP1750-20 BOHLP1750-30
Temperature probes, for agressive liquids, BOLA
Temperatuursondes
Used for temperature measurement in aggressive liquids and cable provides flexible connection from measuring device to medium.
  • Thermocouple in a PTFE-encapsulated stainless steel tube
  • Temperature probe with tip Ø 6 mm and collar ring Ø 12 mm
  • With white PTFE-coated cable (length of 1,5 m, 4 strands)
  • Typical response times of T50 is 7 to 12 seconds and T90 is 14 to 16 seconds
Certificaten: FDA conformed