SP Bel-Art bench-top biohazard bags
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F131660000 F131660001
129-4922EA 53.4 EUR
129-4922 BELAF131660001
SP Bel-Art bench-top biohazard bags
Zakken Afvalzakken
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
  • Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
  • Steam autoclavable at 121 °C
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (129-4948) and Cover (129-1475).