Solvent-based paint pen markers, Bel-Art Products, a part of SP
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F133770000 F133770001
BELAF133770000EA 292 EUR
BELAF133770000 BELAF133770001
Solvent-based paint pen markers, Bel-Art Products, a part of SP
Pennen Afwasbare pennen
Writes on all plastics, glass, metal, wood, paper and cloth, including Teflon® fluoropolymer resins.
  • Precision seal valve provides smooth ink flow and preserves the pen’s life, even if left uncapped
  • When dry, the solvent based ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
  • Markings are insoluble in water or hydrocarbons
  • Resists many solvents, but is easily removed with alcohol or acetone