Test tube half racks, No-Wire™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F187480013 F187480025 F187480016 F187480020 F187480030
212-6150EA 58.7 EUR
212-6150 212-6153 212-6151 212-6152 212-6154
Test tube half racks, No-Wire™, SP Bel-Art
Rekken Tube Racks
PP, white
  • Steam autoclavable at 121 °C
  • Available in 5 grid sizes to accommodate full range of laboratory test tubes
These compact size plastic racks save space in water baths, laminar flow hoods and fume hoods.