Dewar flasks, flat bottom, type FB
Leverancier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1151 1153 1156 1157 1155 1152
478-4501EA 480 EUR
478-4501 KGWA1153 KGWA1156 KGWA1157 KGWA1155 KGWA1152
Dewar flasks, flat bottom, type FB
Dewars en cryogene vaten
Dewar flasks are made up of borosilicate glass 3,3 and stainless-steel cover. For storage of liquid nitrogen, dry ice and other coolants.
  • Low evaporation rate
  • Easy handling
  • Antimagnetic
  • Protective casing made of stucco aluminium
Area of application: Biotechnology, medicinal technique, laboratory technique, cooling of small samples and can also be used as insulating flask.
Certificaten: Conforms with DIN 12492, ISO 3585.
Order Now
Specificaties
Model FB 0 CAL FB 3 CAL FB 12 CAL FB 18 CAL FB 9 CAL FB 1 CAL
Inhoud 0,15 L 0,45 L 1,2 L 2,1 L 0,85 L 0,25 L
Environmentally Preferable
Dagelijkse verdamping 0,25 L 0,35 L 0,7 L 0,9 L 0,55 L 0,35 L
Totale hoogte 170 mm 220 mm 225 mm 275 mm 220 mm 170 mm
Gewicht, leeg 0,3 kg 0,5 kg 1,2 kg 1,8 kg 0,9 kg 0,4 kg