Magnetic stirring bars, PTFE, octagonal, with pivot ring, Spinbar®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F371090028 F371090010 F371090012 F371090019 F371090020 F371090021 F371090030 F371090033 F371100002 F371100003 F371100058 F371100078 F371100112 F371100158 F371100212 F371101256 F371100138 F371100238 F371100118 F371100178 F371101122 F371101128 F371101218 F371101238 F371102128
BELAF371090028EA 28.1 EUR
BELAF371090028 BELAF371090010 BELAF371090012 BELAF371090019 BELAF371090020 BELAF371090021 BELAF371090030 BELAF371090033 BELAF371100002 BELAF371100003 BELAF371100058 442-9344 BELAF371100112 BELAF371100158 442-9348 BELAF371101256 442-9347 442-9349 BELAF371100118 BELAF371100178 BELAF371101122 BELAF371101128 BELAF371101218 BELAF371101238 BELAF371102128
Magnetic stirring bars, PTFE, octagonal, with pivot ring, Spinbar®, SP Bel-Art
Roerders Magnetische roerstaven
PTFE encapsulated octagonal magnetic stirring bars with integral pivot ring are the most commonly used shape. Their interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars.