Tube end fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F702-12 F702-06 F702-10 F702-18 F702-30 F702-34 F702-38 F702-26 F702-24 F702-22 F702-20 F702-08 F702-42 F702-44 F702-40 F702-36 F702-32 F702-28 F702-02 F702-04
BOHLF702-12EA 16.9 EUR
BOHLF702-12 BOHLF702-06 BOHLF702-10 BOHLF702-18 BOHLF702-30 BOHLF702-34 BOHLF702-38 BOHLF702-26 BOHLF702-24 BOHLF702-22 BOHLF702-20 BOHLF702-08 BOHLF702-42 BOHLF702-44 BOHLF702-40 BOHLF702-36 BOHLF702-32 BOHLF702-28 BOHLF702-02 BOHLF702-04
Tube end fittings, BOLA
Fittingen en connectoren
Tube end fittings with male thread and PA washers.
  • Made of PTFE or PP
  • Temperature resistant from −200 to 260 °C (PTFE) and from −10 to 100 °C (PP)
  • Chemical resistant