Plier decappers, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
224372 224373
WHEA224372EA 228 EUR
WHEA224372 WHEA224373
Plier decappers, WHEATON®
Capsuleer- en decapsuleertangen
These pliers decappers are constructed of steel with grey plastic coated handles. To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal.