Bekers voor medicijnen
Leverancier: BOETTGER PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06.103.0100 06.103.0102 06.103.0104 06.103.0101 06.103.0103
216-4731EA 4.1 EUR
216-4731 BOET06.103.0102 BOET06.103.0104 BOET06.103.0101 BOET06.103.0103
Bekers voor medicijnen
Bekers
PP, translucent.
  • Highly transparent, optimum visibility of the content
  • Pressure-resistant and unbreakable
  • Snap-on cover with writing area available as an option
Universal beaker for a wide variety of uses. Ideal for use as a measuring beaker for medicine or to collect/distribute tablets.