Forceps, ESD-safe
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NK6 NK7 NK2A NKR NK35A
232-2171EA 25.5 EUR
232-2171 232-2172 232-2173 232-2174 232-2175
Forceps, ESD-safe
Pincetten
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
  • Electrically conductive
  • Very high chemical resistance to acids and bases
  • Heat resistant up to 175 °C
DIN 53482
Certificaten: DIN 53482