Cultuurbuizen, glas
Leverancier: RENES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31.00.56 31.00.57 31.00.35
212-0292EA 19.7 EUR
212-0292 212-0293 212-0291
Cultuurbuizen, glas
Buisjes Cultuurbuisjes
Glass, clear.
  • Very good chemical resistance
  • For use in applications without high thermal loads
Round bottom, with rim.