Reinigingsoplossing, Heavy-Duty, MegaClean™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MC1-Q MC1-5G MC1-G
MICRMC1-QEA 36.5 EUR
MICRMC1-Q 115-3180 115-0518
Reinigingsoplossing, Heavy-Duty, MegaClean™
Reinigingsmiddelen Detergenten en oppervlaktereinigers
A non-flammable, heavy-duty cleaner and degreaser that removes grease, adhesives, and photo-resistant stains from all general surfaces. Filtered to 0.1 µm, this cleaner is ideal for semiconductor, electronics, aerospace, and clean-build operations.
  • Non-flammable, heavy-duty cleanroom degreaser
  • Effective at low concentrations
  • Filtered to 0,1 µm
  • Water soluble
The Micronova MegaClean™ heavy-duty cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Available in 1 or 5 gallon bottles.