Reinigingsmiddel voor vloeren, NovaClean™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NC1-G NC1-Q NC1-5G
115-0289EA 186 EUR
115-0289 115-0290 115-2680
Reinigingsmiddel voor vloeren, NovaClean™
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Concentrated, all-purpose cleanroom detergent is designed for use between disinfectant applications and for residue removal. Free of mobile ions, this formula is effective on most surfaces, including conductive flooring and semi conductor applications.
  • All-purpose cleanroom detergent
  • Ideal for pharmaceutical and biotech applications
  • Use NC10-G amine-free formula for semi-conductor applications
  • Anti-static and safe on conductive flooring
  • Yields 60 to 128 gallons (227 to 485 L) of cleaning solution per 1 gallon container
  • Filtered to 0,1 µm; free of mobile