Maalbekers „Comfort“ voor PM 100, PM 100CM, PM 200, PM 400
Leverancier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01.462.0239 01.462.0240
412-0083EA 596 EUR
412-0083 412-0084
Maalbekers „Comfort“ voor PM 100, PM 100CM, PM 200, PM 400
Potjes Potten/kommen voor malen
Effective grinding can be achieved by the selection of grinding jars and grinding balls from a wide range of sizes and materials of construction. The ‘comfort’ grinding jars have several safety and design features that make them easy to use.
  • O-ring for gas-tight and dust-proof seal
  • Stainless steel protective jacket for jars that are not made of steel
  • Gap between flanges of jar and edge of lid for easy opening