MonoBox, TripleBox
Leverancier: DELTA T
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V020019 V020020 V020021 V020014 V020015 V020013
478-9934EA 351 EUR
478-9934 478-9935 478-9936 478-9929 478-9930 478-9928
MonoBox, TripleBox
Dozen Transportdozen
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
  • Eliminates the cost of expensive refrigeration transportation
  • Maintains the required temperature
  • Reusable
The MonoTripleBox can be used to transport test tubes or other products.