MonoTriple Box
Leverancier: DELTA T
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V020017 V020018 V020016
478-9932EA 0 EUR
478-9932 478-9933 478-9931
MonoTriple Box
Dozen Transportdozen
The MonoBox are designed for transporting from one to three TempShells. The cooling element pairs fit inside the foam outer casing and are kept at the correct temperature for up to 36 hours. The MonoBox are intended for transporting bags, specimen or other products.
  • For transporting by road, rail or post
  • For blood between the blood bank and the donor facility, as well as for deliveries of small volume pharmaceutical items
  • Made from two pieces in two different sizes that can be interchanged to create different inner volumes, for either 1, 2 or 3 TempShells
Weight: 700 to 1000 g