Tissue and biopsy cassettes in QuickLoad™Sleeves, Histosette® II
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M486-8SL M486-9SL M485-7SL M485-15SL M485-16SL M485-17SL M485-18SL M486-7SL M485-8SL M485-9SL
720-1517EA 217 EUR
720-1517 720-1518 720-1511 720-1507 720-1508 720-1509 720-1510 720-1516 720-1512 720-1513
Tissue and biopsy cassettes in QuickLoad™Sleeves, Histosette® II
Histologie-cassettes Cassettes voor weefselbehandeling
Made of acetal polymer. Suitable for hoppers accepting plastic sleeves, these cassettes will load in cassette labeling instruments in one simple operation. Save time and money with these convenient stacks of 75 unique cassettes. No more manual insertion, one cassette at a time.
  • Keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Biopsy model designed to hold small biopsy samples securely during the embedding process
  • Anterior printing area is at 45° angle
  • One-piece disposable plastic cover eliminates the need for reusable steel lids. Available in non-cytoxic, non-metallic popular colors
Leveringsinformatie: Each case contains 10 sleeves and 750 covers.