Infusion filters, Sterifix®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4550200 4550250
720-2516EA 81.7 EUR
720-2516 720-2517
Infusion filters, Sterifix®
Infusen
Flexible tube with integrated 5 µm particle filter.
  • Latex-free