Venipuncture sets, Venofix®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4056310N 4056370N 4056337N
BRAU4056310NEA 26.2 EUR
BRAU4056310N BRAU4056370N BRAU4056337N
Venipuncture sets, Venofix®
Canules
Venofix sets with puncture needle made of high-quality chromium-nickel steel and a micro-siliconised precision bevel.
  • Latex and PVC free, Lock fitting, closing cone lock
  • Wings with hand grips for puncturing and fixing
Suitable for short-term injections, infusions or transfusions and blood sampling.
Bestelinformatie: Include a puncture needle, interlocking wings and a 30 cm kink-resistant, flexible, transparent tube.