Precast agarose gels, RunOne™
Leverancier: EMBITEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GE-4700 GE-4707 GE-4540 GE-4543 GE-4652 GE-3570 GE-4570 GE-4572 GE-3640 GE-3642 GE-4640 GE-4642 GE-3670 GE-3672 GE-4670 GE-4672
730-0409EA 111 EUR
730-0409 730-0410 730-0395 EMBIGE-4543 730-0406 730-0374 730-0398 730-0399 730-0377 730-0378 730-0402 730-0403 730-0381 730-0382 730-0407 730-0408
Precast agarose gels, RunOne™
Gel voor elektroforese Voorgegoten gels
RunOne™ precast gels do not require a specific gel apparatus but are optimised for use with the RunOne™ Electrophoresis System. A choice of 100, 104 or 108 well formats make these gels convenient for all screening applications.
  • Gel format matches that of standard 96-well PCR plates avoiding confusion over results
  • Gel is exposed to buffer on both ends resulting in efficient running and less gel distortion
  • UV transmissible gel trays make gels easy to handle and view
  • Multi-channel compatible wells allow fast and easy gel loading in multiples of 8 or 12
  • Short run times - 96 samples plus markers can run in under 20 minutes