VWR®, zelfklevende PCR-filmafdichtingen
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
391-1254EA 98.3 EUR
391-1254 391-1255 391-1294 731-0321
VWR®, zelfklevende PCR-filmafdichtingen
Afdichtingen Afdichtingen van microtiterplaten
These heat resistant 74,9 µm thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For real-time PCR applications, where maximum optical clarity is required, use optically-clear polyester films.
  • Aanbevolen voor temperaturen van −40 tot +120 °C
  • Gecertificeerd DNase-, RNase- en nucleasevrij
Elke folie l×b: 135,1×79,4 mm met voldoende afdichtend oppervlak voor alle PCR-platen.