qScript™ One-Step qRT-PCR kits
Leverancier: Quantabio
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
qScript™
733-1261EA 908 EUR
733-1261 733-1259 733-1257
qScript™ One-Step qRT-PCR kits
Reagentia voor nucleïnezuren qPCR/RT-qPCR Enzymes and Kits
Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures. These kits are optimised to deliver maximum RT-PCR efficiency and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. As a result the qScript™ One-Step qRT-PCR Kits offer improved sensitivity over other commercially available qPCR reagents. The qScript™ One-Step Master Mix maximises the activities of both reverse transcriptase and PCR enzymes, while minimising primer-dimer formation. High specificity is assured with AccuStart™ Taq DNA Polymerase, which is bound by monoclonal antibodies for superior hot-start. Upon heating to 95 °C, the antibodies denature irreversibly releasing fully active and unmodified Taq DNA polymerase. qScript™ One-Step qRT-PCR Kits are available for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye.
  • Groot dynamisch bereik zorgt voor betrouwbare gegevens
  • Uitermate gevoelig voor RNA-kwantificering met beperkt aantal kopieën
  • Zuivere duplexamplificatie voor nauwkeurigere resultaten
  • Eenvoudig, snel protocol met minder variabiliteit