WHEATON® Premium Rubber Stoppers
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W224100-406 W224100-421 W224100-408 W224100-400 W224100-420 W224100-405 W224100-402
548-2586EA 710 EUR
548-2586 WHEAW224100-421 WHEAW224100-408 548-2585 WHEAW224100-420 548-2631 WHEAW224100-402
WHEATON® Premium Rubber Stoppers
Stoppen Rubberen stoppen
Ultra-pure rubber stoppers made from a bromobutyl rubber compound containing the highest level of chemical purity. These stoppers demonstrate very low levels of moisture absorption and extractables.
  • Compatible with Water For Injection (WFI) applications
  • Free from natural rubber or natural rubber latex
They are ideal for use in most lyophilisation applications. Complete coat rubber stoppers are completely coated with a fluorinated polymer that has barrier properties similar to PTFE.
The fluorinated polymer coating is all that comes in contact withMore Product Information
Order Now
Specificaties
Environmentally Preferable