Control valves, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E672-54 E672-56
BOHLE672-54EA 538 EUR
BOHLE672-54 BOHLE672-56
Control valves, BOLA
Ventielen Luchtregelventielen
Body made of PTFE with 2 connections and nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Volume flow can be regulated manually
  • Suitable for panel mounting
  • Easy disassembly for cleaning
  • Universal chemical resistance, the flowing product is only exposed to PTFE