Control valves with GL thread, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E694-18 E694-14
BOHLE694-18EA 810 EUR
BOHLE694-18 BOHLE694-14
Control valves with GL thread, BOLA
Ventielen Luchtregelventielen
Two-way valve with straight bore and two connections with GL thread completely made of PTFE.
  • Suitable for vacuum
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Applications include for distributing liquids or gases, manual regulation for constant flow.