Manifold blocks, miniature, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F712-12 F712-10
BOHLF712-12EA 306 EUR
BOHLF712-12 BOHLF712-10
Manifold blocks, miniature, BOLA
Spruitstukken
Block made of PTFE with one inlet and four outlets with female thread UNF ¼" 28 G.
  • Universal chemical resistance
  • Available with four stopcocks