Spray bottles, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
226436 226434 226433
WHEA226436EA 618 EUR
WHEA226436 WHEA226434 WHEA226433
Spray bottles, WHEATON®
Flessen Flessen met verstuiver
Bottle made of white LDPE (Low Density Polyethylene).
  • Includes bottle, atomizer tip, dip tube and screw cap
  • Screw cap made from polypropylene
Oval spray bottle.