Stopcocks with leak-proof plug, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F308910000 F308900000 F308940000 F308950000
229-7436EA 102 EUR
229-7436 229-7435 229-7437 229-7438
Stopcocks with leak-proof plug, SP Bel-Art
Afsluitkranen
Stopcocks with a removable, leak-proof plug made of chemical-resistant, non-stick PTFE.
  • Instant cut-off control
  • Pressure rating is 240 KPa (35 psi)
  • Very good chemical and temperature resistance